CAS RN 84-67-3
IUPAC Name:
2,2'-dimethyl-[1,1'-biphenyl]-4,4'-diamine
Synonyms:
2,2'-dimethyl-[1,1'-biphenyl]-4,4'-diamine
Molecular Weight (g/mol):
212.3
Molecular Formula:
C14H16N2
InChi Key:
QYIMZXITLDTULQ-UHFFFAOYSA-N
SMILES:
CC1=CC(N)=CC=C1C1=CC=C(N)C=C1C
1
–
1
of
1
results
1
m-Tolidine 98.0+%, TCI America™
CAS: 84-67-3 Molecular Formula: C14H16N2 Molecular Weight (g/mol): 212.30 MDL Number: MFCD02269175 InChI Key: QYIMZXITLDTULQ-UHFFFAOYSA-N Synonym: 2,2′C-Dimethylbenzidine, 4,4′C-Diamino-2,2′C-dimethylbiphenyl PubChem CID: 66537 IUPAC Name: 2,2'-dimethyl-[1,1'-biphenyl]-4,4'-diamine SMILES: CC1=CC(N)=CC=C1C1=CC=C(N)C=C1C