missing translation for 'onlineSavingsMsg'
Learn More

m-Tolidine 98.0+%, TCI America™

Catalog No. T309825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T309825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T309825G Supplier TCI America Supplier No. T309825G
Only null left

Chemical Identifiers

CAS 84-67-3
Molecular Formula C14H16N2
Molecular Weight (g/mol) 212.30
MDL Number MFCD02269175
InChI Key QYIMZXITLDTULQ-UHFFFAOYSA-N
Synonym 2,2′C-Dimethylbenzidine, 4,4′C-Diamino-2,2′C-dimethylbiphenyl
PubChem CID 66537
IUPAC Name 2,2'-dimethyl-[1,1'-biphenyl]-4,4'-diamine
SMILES CC1=CC(N)=CC=C1C1=CC=C(N)C=C1C

Specifications

Melting Point 106°C
Quantity 25 g
Formula Weight 212.30
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material m-Tolidine
EINECSNumber (4)-0803
TSCA No