accessibility menu, dialog, popup

UserName

CAS RN 84-26-4

IUPAC Name: 3,13,21-triazapentacyclo[11.8.0.0²,¹⁰.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2(10),4,6,8,15,17,19-octaen-14-one
Synonyms: rutaecarpine rutecarpine (8CI) rutacarpine rutaecarpin rutecarpine rhetine
Molecular Weight (g/mol): 287.32
Molecular Formula: C18H13N3O
InChi Key: ACVGWSKVRYFWRP-UHFFFAOYSA-N
SMILES: O=C1N2CCC3=C(NC4=CC=CC=C34)C2=NC2=CC=CC=C12
Percent Purity 
  • (1)
  • (1)
Grade 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results