missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rutaecarpine

Catalog No. AC465982500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465982500 250 mg
AC465980010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465982500 Supplier Thermo Scientific Chemicals Supplier No. 465982500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 84-26-4 | C18H13N3O, C18H13N3O | 287.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 84-26-4
Molecular Formula C18H13N3O

Specifications

Quantity 250 mg
Solubility Solubility aq. soln.: pracitically insoluble. Other solubilities: soluble in alcohol, benzene, chloroform, ether
Melting Point 259.0°C to 260.0°C
Formula Weight 287.32
Chemical Name or Material Rutaecarpine

RUO – Research Use Only