CAS RN 83915-83-7
CAS RN 83915-83-7
IUPAC Name:
(2S)-1-[(2S)-6-amino-2-{[(1S)-1-carboxy-3-phenylpropyl]amino}hexanoyl]pyrrolidine-2-carboxylic acid dihydrate
Synonyms:
lisinopril dihydrate renacor
Molecular Weight (g/mol):
441.53
Molecular Formula:
C21H35N3O7
InChi Key:
CZRQXSDBMCMPNJ-ZUIPZQNBSA-N
SMILES:
O.O.NCCCC[C@H](N[C@@H](CCC1=CC=CC=C1)C(O)=O)C(=O)N1CCC[C@H]1C(O)=O
1
–
1
of
1
results
1
Lisinopril Dihydrate 98.0+%, TCI Americaâ„¢
CAS: 83915-83-7 Molecular Formula: C21H33N3O6 Molecular Weight (g/mol): 423.51 MDL Number: MFCD01698825 InChI Key: BAVDEDVBIHTHJQ-UVJOBNTFSA-N Synonym: 1-[Nalpha-[(S)-1-Carboxy-3-phenylpropyl]-L-lysyl]-L-proline PubChem CID: 22887897 IUPAC Name: (2S)-1-[(2S)-6-amino-2-[[(1S)-1-carboxy-3-phenylpropyl]amino]hexanoyl]pyrrolidine-2-carboxylic acid;hydrate SMILES: C1CC(N(C1)C(=O)C(CCCCN)NC(CCC2=CC=CC=C2)C(=O)O)C(=O)O.O