missing translation for 'onlineSavingsMsg'
Learn More

Lisinopril Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015658
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015658 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015658 Supplier Merck Emd Millipore Supplier No. PHR11431G
Only null left

Lisinopril is a lysine derivative of enalaprilat and belongs to the group of angiotensin converting enzyme (ACE) inhibitors. It is widely used as an orally active medication in the treatment of high blood pressure and may be used to treat congestive heart failure.

Pharmaceutical (small molecule)

Specifications

CAS 83915-83-7
Physical Form Neat
Linear Formula C21H31O5N3 · 2H2 O
MDL Number MFCD01698825
Synonym (S)-1-[N2-(1-carboxy-3-phenylpropyl)-lysyl-proline dihydrate; MK-521
Type Certified Reference Material
Quantity 1 g
Formula Weight 441.52
Grade Certified Reference Material