CAS RN 83792-47-6
CAS RN 83792-47-6
IUPAC Name:
5-{[amino(4-methylbenzenesulfonamido)methylidene]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoic acid
Synonyms:
5-{[amino(4-methylbenzenesulfonamido)methylidene]amino}-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}pentanoic acid
Molecular Weight (g/mol):
550.63
Molecular Formula:
C28H30N4O6S
InChi Key:
JRRARHJPRLAGNT-UHFFFAOYNA-N
SMILES:
CC1=CC=C(C=C1)S(=O)(=O)NC(N)=NCCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O