missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nomega-tosyl-L-arginine 98.0+%, TCI America™

Catalog No. F08831G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F08831G Supplier TCI America Supplier No. F08831G
Only null left

Specifications

CAS 83792-47-6
Melting Point 94°C
Color Yellow
Molecular Formula C28H30N4O6S
Quantity 1 g
MDL Number MFCD00063331
Synonym fmoc-arg tos-oh, nalpha-fmoc-nomega-tosyl-l-arginine, nalpha-9h-fluoren-9-ylmethoxy carbonyl-nomega-tosyl-l-arginine, fmoc-arginine tos, fmoc-l-arg tos-oh, arginine derivative 6, pubchem18947, fmoc-arg tos-oh et2o, fmoc-n-gamma-tosyl-l-arginine etherate, fmoc-nw-4-toluenesulfonyl-l-arginine
InChI Key JRRARHJPRLAGNT-VWLOTQADSA-N
SMILES CC1=CC=C(C=C1)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)N
IUPAC Name (2S)-5-[[amino-[(4-methylphenyl)sulfonylamino]methylidene]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid
Molecular Weight (g/mol) 550.63
PubChem CID 3010412
Formula Weight 550.63
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nomega-tosyl-L-arginine
Show More Show Less
TSCA No
Recommended Storage Refrigerator