accessibility menu, dialog, popup

UserName

CAS RN 83781-47-9

IUPAC Name: trimethyl({2-[(trimethylsilyl)methyl]phenyl}methyl)azanium iodide
Synonyms: trimethyl({2-[(trimethylsilyl)methyl]phenyl}methyl)azanium iodide
Molecular Weight (g/mol): 363.36
Molecular Formula: C14H26INSi
InChi Key: GTFSUBIVRHGESQ-UHFFFAOYSA-M
SMILES: [I-].C[N+](C)(C)CC1=CC=CC=C1C[Si](C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results