missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl[2-[(trimethylsilyl)methyl]benzyl]ammonium Iodide 98.0+%, TCI America™

Catalog No. T26255G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T26255G Supplier TCI America Supplier No. T26255G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 83781-47-9
Molecular Formula C14H26INSi
Molecular Weight (g/mol) 363.358
InChI Key GTFSUBIVRHGESQ-UHFFFAOYSA-M
PubChem CID 12035585
IUPAC Name trimethyl-[[2-(trimethylsilylmethyl)phenyl]methyl]azanium;iodide
SMILES C[N+](C)(C)CC1=CC=CC=C1C[Si](C)(C)C.[I-]

Specifications

Color White
Quantity 5 g
Formula Weight 363.36
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trimethyl[2-[(trimethylsilyl)methyl]benzyl]ammonium Iodide