CAS RN 835-11-0
IUPAC Name:
2-(2-hydroxybenzoyl)phenol
Synonyms:
2,2'-dihydroxybenzophenone
Molecular Weight (g/mol):
214.22
Molecular Formula:
C13H10O3
InChi Key:
YIYBRXKMQFDHSM-UHFFFAOYSA-N
SMILES:
OC1=CC=CC=C1C(=O)C1=CC=CC=C1O
1
–
1
of
1
results
1
2,2'-Dihydroxybenzophenone 99.0+%, TCI America™
CAS: 835-11-0 Molecular Formula: C13H10O3 Molecular Weight (g/mol): 214.22 MDL Number: MFCD00002217 InChI Key: YIYBRXKMQFDHSM-UHFFFAOYSA-N PubChem CID: 70038 ChEBI: CHEBI:34209 IUPAC Name: bis(2-hydroxyphenyl)methanone SMILES: C1=CC=C(C(=C1)C(=O)C2=CC=CC=C2O)O