missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dihydroxybenzophenone 99.0+%, TCI America™

Catalog No. D109925G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D109925G 25 g
D109910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D109925G Supplier TCI America Supplier No. D109925G
Only null left

Chemical Identifiers

CAS 835-11-0
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00002217
InChI Key YIYBRXKMQFDHSM-UHFFFAOYSA-N
PubChem CID 70038
ChEBI CHEBI:34209
IUPAC Name bis(2-hydroxyphenyl)methanone
SMILES C1=CC=C(C(=C1)C(=O)C2=CC=CC=C2O)O

Specifications

Melting Point 63°C
Color Yellow
Quantity 25 g
Formula Weight 214.22
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Dihydroxybenzophenone