CAS RN 83482-57-9
IUPAC Name:
4-[4-(2-phenylpropan-2-yl)phenoxy]benzene-1,2-dicarbonitrile
Synonyms:
4-[4-(2-phenylpropan-2-yl)phenoxy]benzene-1,2-dicarbonitrile
Molecular Weight (g/mol):
338.41
Molecular Formula:
C23H18N2O
InChi Key:
IANCXVAWMHHHKK-UHFFFAOYSA-N
SMILES:
CC(C)(C1=CC=CC=C1)C1=CC=C(OC2=CC(C#N)=C(C=C2)C#N)C=C1
1
–
1
of
1
results
1
4-(4-alpha-Cumylphenoxy)phthalonitrile 98.0+%, TCI America™
CAS: 83482-57-9 Molecular Formula: C23H18N2O Molecular Weight (g/mol): 338.41 MDL Number: MFCD00059048 InChI Key: IANCXVAWMHHHKK-UHFFFAOYSA-N Synonym: 2-[4-(3,4-Dicyanophenoxy)phenyl]-2-phenylpropane PubChem CID: 371798 IUPAC Name: 4-[4-(2-phenylpropan-2-yl)phenoxy]benzene-1,2-dicarbonitrile SMILES: CC(C)(C1=CC=CC=C1)C1=CC=C(OC2=CC(C#N)=C(C=C2)C#N)C=C1