missing translation for 'onlineSavingsMsg'
Learn More

4-(4-alpha-Cumylphenoxy)phthalonitrile 98.0+%, TCI America™

Catalog No. C11635G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C11635G 5 g
C11631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C11635G Supplier TCI America Supplier No. C11635G

Chemical Identifiers

CAS 83482-57-9
Molecular Formula C23H18N2O
Molecular Weight (g/mol) 338.41
MDL Number MFCD00059048
InChI Key IANCXVAWMHHHKK-UHFFFAOYSA-N
Synonym 2-[4-(3,4-Dicyanophenoxy)phenyl]-2-phenylpropane
PubChem CID 371798
IUPAC Name 4-[4-(2-phenylpropan-2-yl)phenoxy]benzene-1,2-dicarbonitrile
SMILES CC(C)(C1=CC=CC=C1)C1=CC=C(OC2=CC(C#N)=C(C=C2)C#N)C=C1

Specifications

Melting Point 91°C
Color White-Yellow
Quantity 5 g
Formula Weight 338.41
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-alpha-Cumylphenoxy)phthalonitrile