accessibility menu, dialog, popup

UserName

CAS RN 833-81-8

IUPAC Name: [(1E)-2-phenylprop-1-en-1-yl]benzene
Synonyms: [(1E)-2-phenylprop-1-en-1-yl]benzene
Molecular Weight (g/mol): 194.28
Molecular Formula: C15H14
InChi Key: OVZXISBUYCEVEV-OUKQBFOZSA-N
SMILES: C\C(=C/C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results