accessibility menu, dialog, popup

UserName

NH2NNNHNBr

CAS RN 83255-86-1

IUPAC Name: 3-bromo-2H-pyrazolo[3,4-d]pyrimidin-4-amine
Synonyms: 3-bromo-2H-pyrazolo[3,4-d]pyrimidin-4-amine
Molecular Weight (g/mol): 214.03
Molecular Formula: C5H4BrN5
InChi Key: GZQVGSRUUTUJNG-UHFFFAOYSA-N
SMILES: NC1=NC=NC2=NNC(Br)=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results