missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-1H-pyrazolo[3,4-d]pyrimidin-4-amine 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 350°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 214.03 |
| Percent Purity | ≥97.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Bromo-1H-pyrazolo[3,4-d]pyrimidin-4-amine |
Safety and Handling
| TSCA | No |