CAS RN 83048-65-1
CAS RN 83048-65-1
IUPAC Name:
(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)trimethoxysilane
Synonyms:
(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)trimethoxysilane
Molecular Weight (g/mol):
568.3
Molecular Formula:
C13H13F17O3Si
InChi Key:
HJIMAFKWSKZMBK-UHFFFAOYSA-N
SMILES:
CO[Si](CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)(OC)OC
1
–
1
of
1
results
1
Trimethoxy(1H,1H,2H,2H-heptadecafluorodecyl)silane 98.0+%, TCI America™
CAS: 83048-65-1 Molecular Formula: C13H13F17O3Si Molecular Weight (g/mol): 568.30 MDL Number: MFCD07368748 InChI Key: HJIMAFKWSKZMBK-UHFFFAOYSA-N Synonym: Trimethoxy(1H,1H,2H,2H-perfluorodecyl)silane, 1H,1H,2H,2H-Heptadecafluorodecyltrimethoxysilane