missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethoxy(1H,1H,2H,2H-heptadecafluorodecyl)silane 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 80°C |
| Quantity | 5 g |
| Formula Weight | 568.30 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Trimethoxy(1H,1H,2H,2H-heptadecafluorodecyl)silane |
Safety and Handling
| TSCA | Yes |