accessibility menu, dialog, popup

UserName

CAS RN 830-96-6

IUPAC Name: 3-(1H-indol-3-yl)propanoic acid
Synonyms: indolylpropionic acid β-indole-3-propionate ipa (auxin) β-indole-3-propionic acid
Molecular Weight (g/mol): 189.21
Molecular Formula: C11H11NO2
InChi Key: GOLXRNDWAUTYKT-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CNC2=CC=CC=C12
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)