accessibility menu, dialog, popup

UserName

CAS RN 83-79-4

IUPAC Name: (1S,6R,13S)-16,17-dimethoxy-6-(prop-1-en-2-yl)-2,7,20-trioxapentacyclo[11.8.0.0³,¹¹.0⁴,⁸.0¹⁴,¹⁹]henicosa-3,8,10,14(19),15,17-hexaen-12-one
Synonyms: cube root caswell no. 725 chem-mite ronone haiari 5'β-rotenone deril canex pro-nox fish curex flea duster
Molecular Weight (g/mol): 394.42
Molecular Formula: C23H22O6
InChi Key: JUVIOZPCNVVQFO-HBGVWJBISA-N
SMILES: COC1=CC2=C(C=C1OC)[C@H]1[C@@H](CO2)OC2=C3C[C@@H](OC3=CC=C2C1=O)C(C)=C
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results