missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Rotenone

Catalog No. 5573681GM
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5573681GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5573681GM Supplier MilliporeSigma™ Supplier No. 5573681GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A mitochondrial toxin and a potent, reversible, and competitive inhibitor of complex I (NADH-CoQ reductase) of the respiratory chain.

Packaged Under Inert Gas

Specifications

Color White to Off-white
Description 98% by HPLC
Product Type Inhibitor
Target Proteins NADH-CoQ reductase
CAS 83-79-4
Molecular Formula C23H22O6
Formula Weight 394.4g/mol
Solubility CHCl3, DMSO, or EtOH
Content And Storage Protect from light
Form Solid
Quantity 1 g
Show More Show Less