accessibility menu, dialog, popup

UserName

CAS RN 82957-06-0

CH3SOOHONH2H2NO

CAS RN 82957-06-0

IUPAC Name: 6-(diaminomethylidene)-2,6-dihydronaphthalen-2-one; methanesulfonic acid
Synonyms: 6-(diaminomethylidene)naphthalen-2-one; methanesulfonic acid
Molecular Weight (g/mol): 282.31
Molecular Formula: C12H14N2O4S
InChi Key: OHFQGSXMJXLUAG-UHFFFAOYSA-N
SMILES: CS(O)(=O)=O.NC(N)=C1C=CC2=CC(=O)C=CC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results