missing translation for 'onlineSavingsMsg'
Learn More

6-Amidino-2-naphthol Methanesulfonate 99.0+%, TCI America™

Catalog No. A11935G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11935G 5 g
A11931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A11935G Supplier TCI America Supplier No. A11935G

Chemical Identifiers

CAS 82957-06-0
Molecular Formula C12H14N2O4S
Molecular Weight (g/mol) 282.31
MDL Number MFCD00143493
InChI Key OHFQGSXMJXLUAG-UHFFFAOYSA-N
Synonym 6-Amidino-2-naphthol Methanesulfonic Acid, 6-Hydroxynaphthalene-2-carboximidamide Methanesulfonate, 6-Carbamimidoyl-2-naphthol Methanesulfonate
PubChem CID 12396152
IUPAC Name 6-(diaminomethylidene)-2,6-dihydronaphthalen-2-one; methanesulfonic acid
SMILES CS(O)(=O)=O.NC(N)=C1C=CC2=CC(=O)C=CC2=C1

Specifications

Quantity 5 g
Formula Weight 282.31
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Amidino-2-naphthol Methanesulfonate