accessibility menu, dialog, popup

UserName

CH3OO(S)NH(S)H3COHO

CAS RN 82717-96-2

IUPAC Name: (2S)-2-{[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino}propanoic acid
Synonyms: (2S)-2-{[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino}propanoic acid
Molecular Weight (g/mol): 279.34
Molecular Formula: C15H21NO4
InChi Key: CEIWXEQZZZHLDM-AAEUAGOBSA-N
SMILES: CCOC(=O)[C@H](CCC1=CC=CC=C1)N[C@@H](C)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results