missing translation for 'onlineSavingsMsg'
Learn More

N-[(S)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine 98.0+%, TCI America™

Catalog No. E05305G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05305G 5 g
E053025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E05305G Supplier TCI America Supplier No. E05305G
Only null left

Chemical Identifiers

CAS 82717-96-2
Molecular Formula C15H21NO4
Molecular Weight (g/mol) 279.34
MDL Number MFCD00209976
InChI Key CEIWXEQZZZHLDM-AAEUAGOBSA-N
Synonym s-2-s-1-ethoxy-1-oxo-4-phenylbutan-2-yl amino propanoic acid, n-s-1-ethoxycarbonyl-3-phenylpropyl-l-alanine, unii-10sz4ij77b, n-1 s-ethoxycarbonyl-3-phenylpropyl-l-alanine, n-s-+-1-ethoxycarbonyl-3-phenylpropyl-l-alanine, 2s-2-2s-1-ethoxy-1-oxo-4-phenylbutan-2-yl amino propanoic acid, s,s-n-ethoxy carbonyl-3-phenylpropyl alanine, n-s-1-carboxyethyl-l-homophenylalanine ethyl ester, s,s-n-1-ethoxycarbonyl-3-phenylpropyl alanine
PubChem CID 5702571
IUPAC Name (2S)-2-{[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino}propanoic acid
SMILES CCOC(=O)[C@H](CCC1=CC=CC=C1)N[C@@H](C)C(O)=O

Specifications

Melting Point 150°C
Color White
Quantity 5 g
Formula Weight 279.34
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(S)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine
EINECSNumber (3)-3931
TSCA No