accessibility menu, dialog, popup

UserName

CAS RN 827-43-0

CH3NHN

CAS RN 827-43-0

IUPAC Name: 5-methyl-2-phenyl-1H-imidazole
Synonyms: 4-methyl-2-phenyl-3H-imidazole
Molecular Weight (g/mol): 158.2
Molecular Formula: C10H10N2
InChi Key: TYOXIFXYEIILLY-UHFFFAOYSA-N
SMILES: CC1=CN=C(N1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results