accessibility menu, dialog, popup

UserName

NH2BrBrNOO

CAS RN 827-23-6

IUPAC Name: 2,4-dibromo-6-nitroaniline
Synonyms: 2,4-dibromo-6-nitroaniline
Molecular Weight (g/mol): 295.92
Molecular Formula: C6H4Br2N2O2
InChi Key: OBTUHOVIVLDLLD-UHFFFAOYSA-N
SMILES: NC1=C(Br)C=C(Br)C=C1[N+]([O-])=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results