missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromo-6-nitroaniline 98.0+%, TCI America™

Catalog No. D22515G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D22515G Supplier TCI America Supplier No. D22515G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 827-23-6
Molecular Formula C6H4Br2N2O2
Molecular Weight (g/mol) 295.918
MDL Number MFCD00012107
InChI Key OBTUHOVIVLDLLD-UHFFFAOYSA-N
Synonym benzenamine, 2,4-dibromo-6-nitro, 2,4-dibromo-6-nitrophenylamine, 2,4-dibromo-6-nitrobenzenamine, acmc-209pq0, 2-nitro-4,6-dibromoaniline, 4,6-dibromo-2-nitroaniline, 2,4-dibromo-6-nitro-aniline, 2,4-dibromo-6-nitro-phenylamine, aniline, 2,4-dibromo-6-nitro, benzenamine,2,4-dibromo-6-nitro
PubChem CID 70009
IUPAC Name 2,4-dibromo-6-nitroaniline
SMILES C1=C(C=C(C(=C1Br)N)[N+](=O)[O-])Br

Specifications

Melting Point 127°C
Quantity 5 g
Formula Weight 295.92
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dibromo-6-nitroaniline