accessibility menu, dialog, popup

UserName

CAS RN 827-15-6

IUPAC Name: 1,2,3,4,5-pentafluoro-6-iodobenzene
Synonyms: iodopentafluorobenzene
Molecular Weight (g/mol): 293.96
Molecular Formula: C6F5I
InChi Key: OPYHNLNYCRZOGY-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(I)C(F)=C1F
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)