missing translation for 'onlineSavingsMsg'
Learn More

Pentafluoroiodobenzene (stabilized with Copper chip) 99.0+%, TCI America™

Catalog No. P11885G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11885G 5 g
P118825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11885G Supplier TCI America Supplier No. P11885G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 827-15-6
Molecular Formula C6F5I
Molecular Weight (g/mol) 293.962
MDL Number MFCD00001032
InChI Key OPYHNLNYCRZOGY-UHFFFAOYSA-N
Synonym iodopentafluorobenzene, pentafluoroiodobenzene, benzene, pentafluoroiodo, 2,3,4,5,6-pentafluoroiodobenzene, iodoperfluorobenzene, pentafluorophenyl iodide, 1,2,3,4,5-pentafluoro-6-iodo-benzene, perfluoroiodobenzene, ibf, pubchem19982
PubChem CID 70008
IUPAC Name 1,2,3,4,5-pentafluoro-6-iodobenzene
SMILES C1(=C(C(=C(C(=C1F)F)I)F)F)F

Specifications

Color Yellow
Boiling Point 162°C
Quantity 5 g
Formula Weight 293.96
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Pentafluoroiodobenzene (stabilized with Copper chip)