missing translation for 'onlineSavingsMsg'
Learn More

Raloxifene Hydrochloride 98.0+%, TCI America™

Catalog No. R01095G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R01095G 5 g
R01091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R01095G Supplier TCI America Supplier No. R01095G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 82640-04-8
Molecular Formula C28H28ClNO4S
Molecular Weight (g/mol) 510.045
MDL Number MFCD01938233
InChI Key BKXVVCILCIUCLG-UHFFFAOYSA-N
Synonym raloxifene hydrochloride, raloxifene hcl, evista, keoxifene hydrochloride, 6-hydroxy-2-4-hydroxyphenyl benzo b thiophen-3-yl 4-2-piperidin-1-yl ethoxy phenyl methanone hydrochloride, keoxifene, unii-4f86w47br6, evista raloxifene hydrochloride, 6-hydroxy-2-p-hydroxyphenyl benzo b thien-3-yl-p-2-piperidinoethoxy phenyl ketone, hydrochloride
PubChem CID 54900
ChEBI CHEBI:50740
IUPAC Name [6-hydroxy-2-(4-hydroxyphenyl)-1-benzothiophen-3-yl]-[4-(2-piperidin-1-ylethoxy)phenyl]methanone;hydrochloride
SMILES C1CCN(CC1)CCOC2=CC=C(C=C2)C(=O)C3=C(SC4=C3C=CC(=C4)O)C5=CC=C(C=C5)O.Cl

Specifications

Melting Point 263°C
Quantity 5 g
Formula Weight 510.05
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Raloxifene Hydrochloride
RTECSNumber PC4956925
TSCA No