accessibility menu, dialog, popup

UserName

CAS RN 82575-69-7

IUPAC Name: 4-(4-pentylcyclohexyl)phenol
Synonyms: 4-(4-pentylcyclohexyl)phenol
Molecular Weight (g/mol): 246.39
Molecular Formula: C17H26O
InChi Key: QRAZRBGYBYIGRL-UHFFFAOYSA-N
SMILES: CCCCCC1CCC(CC1)C1=CC=C(O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results