missing translation for 'onlineSavingsMsg'
Learn More

4-(trans-4-Amylcyclohexyl)phenol 98.0+%, TCI America™

Catalog No. A182725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A182725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A182725G Supplier TCI America Supplier No. A182725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 82575-69-7
Molecular Formula C17H26O
Molecular Weight (g/mol) 246.39
MDL Number MFCD00673750
InChI Key QRAZRBGYBYIGRL-UHFFFAOYSA-N
Synonym 1-(trans-4-Amylcyclohexyl)-4-hydroxybenzene, 4-(trans-4-Pentylcyclohexyl)phenol, 1-(trans-4-Pentylcyclohexyl)-4-hydroxybenzene
PubChem CID 4593765
IUPAC Name 4-(4-pentylcyclohexyl)phenol
SMILES CCCCCC1CCC(CC1)C1=CC=C(O)C=C1

Specifications

Melting Point 140°C
Color White-Yellow
Quantity 25 g
Formula Weight 246.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(trans-4-Amylcyclohexyl)phenol