accessibility menu, dialog, popup

UserName

H3COOOOCH3

CAS RN 82491-60-9

IUPAC Name: 1,3-dimethyl 2-cyclopentylpropanedioate
Synonyms: 1,3-dimethyl 2-cyclopentylpropanedioate
Molecular Weight (g/mol): 200.23
Molecular Formula: C10H16O4
InChi Key: KCCNEJJMJHDVCN-UHFFFAOYSA-N
SMILES: COC(=O)C(C1CCCC1)C(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results