missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Cyclopentylmalonate 97.0+%, TCI America™

Catalog No. C162125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C162125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C162125G Supplier TCI America Supplier No. C162125G
Only null left

Chemical Identifiers

CAS 82491-60-9
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.234
MDL Number MFCD03844781
InChI Key KCCNEJJMJHDVCN-UHFFFAOYSA-N
Synonym Cyclopentylmalonic Acid Dimethyl Ester
PubChem CID 14831157
IUPAC Name dimethyl 2-cyclopentylpropanedioate
SMILES COC(=O)C(C1CCCC1)C(=O)OC

Specifications

Color Colorless
Quantity 25 g
Formula Weight 200.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Cyclopentylmalonate