accessibility menu, dialog, popup

UserName

CAS RN 824-39-5

IUPAC Name: sodium 2-nitrobenzen-1-olate
Synonyms: sodium 2-nitrophenolate
Molecular Weight (g/mol): 161.09
Molecular Formula: C6H4NNaO3
InChi Key: AXKBOWBNOCUNJL-UHFFFAOYSA-M
SMILES: [Na+].[O-]C1=CC=CC=C1[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results