missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenol Sodium Salt 99.0+%, TCI America™

Catalog No. N0222500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0222500G 500 g
N022225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0222500G Supplier TCI America Supplier No. N0222500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 824-39-5
Molecular Formula C6H4NNaO3
Molecular Weight (g/mol) 161.092
MDL Number MFCD00065178
InChI Key AXKBOWBNOCUNJL-UHFFFAOYSA-M
Synonym Sodium 2-Nitrophenol
PubChem CID 69985
IUPAC Name sodium;2-nitrophenolate
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])[O-].[Na+]

Specifications

Quantity 500 g
Formula Weight 161.09
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitrophenol Sodium Salt
EINECSNumber (3)-0779
RTECSNumber SM4755000
TSCA Yes