accessibility menu, dialog, popup

UserName

CAS RN 823-40-5

IUPAC Name: 2-methylbenzene-1,3-diamine
Synonyms: 2,6-diaminotoluene
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: RLYCRLGLCUXUPO-UHFFFAOYSA-N
SMILES: CC1=C(N)C=CC=C1N
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)