missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diaminotoluene 98.0+%, TCI America™

Catalog No. D012425G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D012425G 25 g
D012410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D012425G Supplier TCI America Supplier No. D012425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 823-40-5
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00007800
InChI Key RLYCRLGLCUXUPO-UHFFFAOYSA-N
Synonym 2,6-diaminotoluene, 2,6-toluenediamine, 1,3-benzenediamine, 2-methyl, toluene-2,6-diamine, 2,6-tolylenediamine, 2,6-toluylenediamine, 2,6-diamino-1-methylbenzene, 2-methyl-1,3-benzenediamine, 2-methyl-1,3-phenylenediamine, 2-methyl-m-phenylenediamine
PubChem CID 13205
ChEBI CHEBI:76288
IUPAC Name 2-methylbenzene-1,3-diamine
SMILES CC1=C(N)C=CC=C1N

Specifications

Melting Point 106°C
Boiling Point 282°C
Quantity 25 g
UN Number 3077
Formula Weight 122.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Diaminotoluene
EINECSNumber (3)-0126
RTECSNumber XS9750000
TSCA Yes