CAS RN 82297-89-0
IUPAC Name:
bromo(4-fluoro-3-methylphenyl)magnesium
Synonyms:
bromo(4-fluoro-3-methylphenyl)magnesium
Molecular Weight (g/mol):
213.33
Molecular Formula:
C7H6BrFMg
InChi Key:
OLSGDQISPRACFY-UHFFFAOYSA-M
SMILES:
CC1=CC([Mg]Br)=CC=C1F
1
–
1
of
1
results
1
Promotions Available
4-Fluoro-3-methylphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific™
4-Fluoro-3-methylphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific™
CAS: 82297-89-0 Molecular Formula: C7H6BrFMg Molecular Weight (g/mol): 213.33 InChI Key: RDVDTHIVHSZVCT-UHFFFAOYSA-M Synonym: 4-fluoro-3-methylphenylmagnesium bromide,olsgdqispracfy-uhfffaoysa-m,bromo 4-fluoro-3-methylphenyl magnesium,3-methyl-4-fluorophenyl magnesium bromide,4-fluoro-3-methylphenyl magnesium bromide,4-fluoro-3-methylphenylmagnesium bromide, 0.50 m in 2-methf,4-fluoro-3-methylphenylmagnesium bromide 0.5 m in tetrahydrofuran,4-fluoro-3-methylphenylmagnesium bromide solution, 1.0 m in thf,grignard reagent PubChem CID: 2778169 IUPAC Name: magnesium;1-fluoro-2-methylbenzene-4-ide;bromide SMILES: CC1=C[C-]=CC=C1F.[Mg+2].[Br-]