missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methylphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific™

Numéro de catalogue. AC431860500
Modifier l'affichage
Click to view available options
Quantité:
50mL
Emballage:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
AC431860500 50mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. AC431860500 Fournisseur Thermo Scientific Chemicals Code fournisseur 431860500

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

 

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 82297-89-0 , 109-99-9
Formule moléculaire C7H6BrFMg
Poids moléculaire (g/mol) 213.33
Clé InChI RDVDTHIVHSZVCT-UHFFFAOYSA-M
Synonyme 4-fluoro-3-methylphenylmagnesium bromide, olsgdqispracfy-uhfffaoysa-m, bromo 4-fluoro-3-methylphenyl magnesium, 3-methyl-4-fluorophenyl magnesium bromide, 4-fluoro-3-methylphenyl magnesium bromide, 4-fluoro-3-methylphenylmagnesium bromide, 0.50 m in 2-methf, 4-fluoro-3-methylphenylmagnesium bromide 0.5 m in tetrahydrofuran, 4-fluoro-3-methylphenylmagnesium bromide solution, 1.0 m in thf, grignard reagent
PubChem CID 2778169
Nom de l’IUPAC magnesium;1-fluoro-2-methylbenzene-4-ide;bromide
SOURIRES CC1=C[C-]=CC=C1F.[Mg+2].[Br-]

Spécifications

Nom ou substance chimique 4-Fluoro-3-methylphenylmagnesium bromide
CAS % minimum 8.53, 87.21
CAS % maximal 12.79, 91.48
Densité 1.0010g/mL
Point d’éclair −26°C
Emballage AcroSeal™ Glass Bottle
Formule linéaire F(CH3)C6H3MgBr
Quantité 50mL
Gravité spécifique 1.001
Poids de formule 213.33
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Reacts violently with water.
May form explosive peroxides.
Suspected of causing cancer.<br/
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

RUO – Research Use Only