accessibility menu, dialog, popup

UserName

CAS RN 822-48-0

IUPAC Name: 3-(chloromethyl)-3-methyloxetane
Synonyms: 3-(chloromethyl)-3-methyloxetane
Molecular Weight (g/mol): 120.58
Molecular Formula: C5H9ClO
InChi Key: MCZYEFODKAZWIH-UHFFFAOYSA-N
SMILES: CC1(CCl)COC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results