missing translation for 'onlineSavingsMsg'
Learn More

3-(Chloromethyl)-3-methyloxetane 98.0+%, TCI America™

Catalog No. C12295G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12295G 5 g
C122925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C12295G Supplier TCI America Supplier No. C12295G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 822-48-0
Molecular Formula C5H9ClO
Molecular Weight (g/mol) 120.58
MDL Number MFCD01709552
InChI Key MCZYEFODKAZWIH-UHFFFAOYSA-N
Synonym 2-(Chloromethyl)-2-methyl-1,3-epoxypropane
PubChem CID 79112
IUPAC Name 3-(chloromethyl)-3-methyloxetane
SMILES CC1(CCl)COC1

Specifications

Color Colorless
Boiling Point 88°C
Quantity 5 g
UN Number 1993
Formula Weight 120.58
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Chloromethyl)-3-methyloxetane
RTECSNumber RQ6828000
TSCA No