accessibility menu, dialog, popup

UserName

H3COCH3O

CAS RN 822-23-1

IUPAC Name: octadecyl acetate
Synonyms: stearyl acetate
Molecular Weight (g/mol): 312.54
Molecular Formula: C20H40O2
InChi Key: OIZXRZCQJDXPFO-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCOC(C)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results