missing translation for 'onlineSavingsMsg'
Learn More

Octadecyl Acetate 97.0+%, TCI America™

Catalog No. A067525G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A067525G 25 g
A0675250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A067525G Supplier TCI America Supplier No. A067525G
Only null left

Chemical Identifiers

CAS 822-23-1
Molecular Formula C20H40O2
Molecular Weight (g/mol) 312.538
MDL Number MFCD00043649
InChI Key OIZXRZCQJDXPFO-UHFFFAOYSA-N
Synonym Acetic Acid Octadecyl Ester, Acetic Acid Stearyl Ester, Stearyl Acetate
PubChem CID 69968
IUPAC Name octadecyl acetate
SMILES CCCCCCCCCCCCCCCCCCOC(=O)C

Specifications

Color White
Quantity 25 g
Formula Weight 312.54
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Octadecyl Acetate
EINECSNumber (2)-0735
TSCA Yes