accessibility menu, dialog, popup

UserName

CAS RN 82159-09-9

IUPAC Name: 2-[(5Z)-5-[(2E)-2-methyl-3-phenylprop-2-en-1-ylidene]-4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl]acetic acid
Synonyms: kinedak epalrestatum ono 2 ono-2
Molecular Weight (g/mol): 319.39
Molecular Formula: C15H13NO3S2
InChi Key: CHNUOJQWGUIOLD-NFZZJPOKSA-N
SMILES: C\C(\C=C1/SC(=S)N(CC(O)=O)C1=O)=C/C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results