accessibility menu, dialog, popup

UserName

CAS RN 821-06-7

IUPAC Name: (2E)-1,4-dibromobut-2-ene
Synonyms: 2-butene, 1,4-dibromo-
Molecular Weight (g/mol): 213.9
Molecular Formula: C4H6Br2
InChi Key: RMXLHIUHKIVPAB-OWOJBTEDSA-N
SMILES: BrC\C=C\CBr
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)