missing translation for 'onlineSavingsMsg'
Learn More

trans-1,4-Dibromo-2-butene 98.0+%, TCI America™

Catalog No. D294425G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D294425G 25 g
D2944250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D294425G Supplier TCI America Supplier No. D294425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 821-06-7
Molecular Formula C4H6Br2
Molecular Weight (g/mol) 213.9
MDL Number MFCD00000249
InChI Key RMXLHIUHKIVPAB-OWOJBTEDSA-N
Synonym 1,4-dibromo-2-butene, trans-1,4-dibromo-2-butene, e-1,4-dibromobut-2-ene, e-1,4-dibromo-2-butene, 1,4-dibromobut-2-ene, trans-1,4-dibromobut-2-ene, 2-butene, 1,4-dibromo, 2-butene, 1,4-dibromo-, 2e, 2e-1,4-dibromobut-2-ene, 1,4-dibromo-2-butylene
PubChem CID 641245
IUPAC Name (E)-1,4-dibromobut-2-ene
SMILES C(C=CCBr)Br

Specifications

Melting Point 54°C
Color White-Yellow
Boiling Point 76°C
Quantity 25 g
UN Number 2923
Formula Weight 213.90
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-1,4-Dibromo-2-butene
RTECSNumber EM4725000
TSCA Yes
Recommended Storage Refrigerator