accessibility menu, dialog, popup

UserName

CH3OOOOCH3HOCH3OOCl

CAS RN 82034-46-6

IUPAC Name: chloromethyl 1-[(ethoxycarbonyl)oxy]-10-hydroxy-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-1-carboxylate
Synonyms: loteprednol
Molecular Weight (g/mol): 466.96
Molecular Formula: C24H31ClO7
InChi Key: DMKSVUSAATWOCU-UHFFFAOYNA-N
SMILES: CCOC(=O)OC1(CCC2C3CCC4=CC(=O)C=CC4(C)C3C(O)CC12C)C(=O)OCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)